2-Methyl-1-(5-methylthiophen-3-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

426027-5G 426027-1G 426027-500MG
77629-244EA 1484.23 USD
77629-244 77642-740 77650-706
2-Methyl-1-(5-methylthiophen-3-yl)propan-1-one ≥95%
2-Methyl-1-(5-methylthiophen-3-yl)propan-1-one

Precaución: For Research Use Only

Formula: 168.256 g/mol
MW: C₉H₁₂OS
Storage Temperature: Ambient
CAS Number: 1480662-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR