Trimethyl(5-methylthiophen-3-yl)silane ≥95%

Supplier: AOBChem USA

Warning

426023-1G 426023-5G 426023-500MG
77638-708EA 531 USD
77638-708 77655-690 77646-758
Trimethyl(5-methylthiophen-3-yl)silane ≥95%
Trimethyl(5-methylthiophen-3-yl)silane

Precaución: For Research Use Only

Formula: C₈H₁₄SSi
Storage Temperature: Ambient
CAS Number: 1877614-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR