4-(Ethylthio)-2-methylthiophene ≥95%

Supplier: AOBChem USA

Warning

426019-5G 426019-25G 426019-1G 426019-500MG
77629-242EA 1484.23 USD
77629-242 77635-796 77642-736 77650-704
4-(Ethylthio)-2-methylthiophene ≥95%
4-(Ethylthio)-2-methylthiophene

Precaución: For Research Use Only

Formula: C₇H₁₀S₂
Storage Temperature: Ambient
CAS Number: 16238-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR