(2-Chloro-5-methylpyridin-3-yl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

425993-5G 425993-1G 425993-25G 425993-500MG
77633-104EA 1441 USD
77633-104 77638-702 77659-270 77650-698
(2-Chloro-5-methylpyridin-3-yl)(cyclopropyl)methanol ≥95%
(2-Chloro-5-methylpyridin-3-yl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 197.661 g/mol
MW: C₁₀H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 3037703-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR