1-(2-Chloro-5-methylpyridin-3-yl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

425989-5G 425989-1G 425989-500MG
77629-236EA 1553.24 USD
77629-236 77638-700 77646-752
1-(2-Chloro-5-methylpyridin-3-yl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Chloro-5-methylpyridin-3-yl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₆ClNO
MW: 213.704 g/mol
Storage Temperature: Ambient
CAS Number: 3037703-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR