1-(2-Chloro-5-methylpyridin-3-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

425988-5G 425988-1G 425988-25G 425988-500MG
77629-234EA 1484.23 USD
77629-234 77638-698 77659-268 77646-750
1-(2-Chloro-5-methylpyridin-3-yl)-2-methylpropan-1-ol ≥95%
1-(2-Chloro-5-methylpyridin-3-yl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 199.677 g/mol
MW: C₁₀H₁₄ClNO
Storage Temperature: Ambient
CAS Number: 3037692-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR