1-(2-Chloro-5-methylpyridin-3-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

425987-5G 425987-25G 425987-1G 425987-500MG
77629-232EA 1615.04 USD
77629-232 77655-684 77642-732 77650-696
1-(2-Chloro-5-methylpyridin-3-yl)propan-1-ol ≥95%
1-(2-Chloro-5-methylpyridin-3-yl)propan-1-ol

Precaución: For Research Use Only

Formula: 185.651 g/mol
MW: C₉H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 3037676-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR