(2-Chloro-5-methylpyridin-3-yl)(cyclobutyl)methanone ≥95%

Supplier: AOBChem USA

Warning

425984-5G 425984-25G 425984-1G 425984-500MG
77633-102EA 1441 USD
77633-102 77659-266 77642-730 77650-694
(2-Chloro-5-methylpyridin-3-yl)(cyclobutyl)methanone ≥95%
(2-Chloro-5-methylpyridin-3-yl)(cyclobutyl)methanone

Precaución: For Research Use Only

Formula: 209.672 g/mol
MW: C₁₁H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 3037682-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR