(2-Chloro-5-methylpyridin-3-yl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

425981-5G 425981-1G 425981-500MG
77629-230EA 1484.23 USD
77629-230 77638-696 77650-692
(2-Chloro-5-methylpyridin-3-yl)(phenyl)methanone ≥95%
(2-Chloro-5-methylpyridin-3-yl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 231.678 g/mol
MW: C₁₃H₁₀ClNO
Storage Temperature: Ambient
CAS Number: 3037705-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR