1-(2-Chloro-5-methylpyridin-3-yl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

425978-5G 425978-1G 425978-25G 425978-500MG
77629-228EA 1823.1 USD
77629-228 77638-694 77655-682 77646-748
1-(2-Chloro-5-methylpyridin-3-yl)-2-methylpropan-1-one ≥95%
1-(2-Chloro-5-methylpyridin-3-yl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: 197.661 g/mol
MW: C₁₀H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 3037704-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR