1-(2-Chloro-5-methylpyridin-3-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

425974-5G 425974-25G 425974-1G 425974-500MG
77629-224EA 1484.23 USD
77629-224 77655-680 77642-726 77646-744
1-(2-Chloro-5-methylpyridin-3-yl)-2,2-difluoroethanone ≥97%
1-(2-Chloro-5-methylpyridin-3-yl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 205.589 g/mol
MW: C₈H₆ClF₂NO
Storage Temperature: Ambient
CAS Number: 3031799-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR