1-(4-(Cyclopentyloxy)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

425795-5G 425795-25G 425795-1G 425795-500MG
77629-218EA 1484.23 USD
77629-218 77655-676 77642-722 77646-738
1-(4-(Cyclopentyloxy)phenyl)-2,2-difluoroethanone ≥95%
1-(4-(Cyclopentyloxy)phenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 240.2458 g/mol
MW: C₁₃H₁₄F₂O₂
Storage Temperature: Ambient
CAS Number: 3031788-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR