(3-(4-Methylpiperazin-1-yl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

42451-5G 42451-1G 42451-25G 42451-500MG
77633-100EA 799 USD
77633-100 77638-690 77659-262 77646-736
(3-(4-Methylpiperazin-1-yl)phenyl)boronic acid ≥97%
(3-(4-Methylpiperazin-1-yl)phenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₇BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13194680
CAS Number: 1139717-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR