Propanoic acid, 2-[(5-chloro-2-pyridinyl)oxy]-2-methyl-, methyl ester ≥97%

Supplier: AOBChem USA

Warning

424426-5G 424426-25G 424426-1G 424426-500MG
77629-216EA 1484.23 USD
77629-216 77635-794 77638-688 77646-734
Propanoic acid, 2-[(5-chloro-2-pyridinyl)oxy]-2-methyl-, methyl ester ≥97%
Propanoic acid, 2-[(5-chloro-2-pyridinyl)oxy]-2-methyl-, methyl ester

Precaución: For Research Use Only

Formula: 229.66 g/mol
MW: C₁₀H₁₂ClNO₃
Storage Temperature: Ambient
CAS Number: 2974861-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR