Methyl 2-[(6-bromo-2-pyridinyl)oxy]-2-methylpropanoate ≥97%

Supplier: AOBChem USA

Warning

424411-5G 424411-25G 424411-1G 424411-500MG
77629-214EA 1484.23 USD
77629-214 77635-792 77642-720 77650-690
Methyl 2-[(6-bromo-2-pyridinyl)oxy]-2-methylpropanoate ≥97%
Methyl 2-[(6-bromo-2-pyridinyl)oxy]-2-methylpropanoate

Precaución: For Research Use Only

Formula: 274.11 g/mol
MW: C₁₀H₁₂BrNO₃
Storage Temperature: Ambient
CAS Number: 1206247-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR