2-[(6-Bromo-2-pyridinyl)oxy]-2-methylpropanoic acid ≥97%

Supplier: AOBChem USA

Warning

424410-5G 424410-25G 424410-1G 424410-500MG
77629-212EA 1484.23 USD
77629-212 77655-674 77642-718 77650-688
2-[(6-Bromo-2-pyridinyl)oxy]-2-methylpropanoic acid ≥97%
2-[(6-Bromo-2-pyridinyl)oxy]-2-methylpropanoic acid

Precaución: For Research Use Only

Formula: 260.08 g/mol
MW: C₉H₁₀BrNO₃
Storage Temperature: Ambient
CAS Number: 1206249-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR