2-[(5-Chloro-2-pyridinyl)oxy]-2-methylpropanoic acid ≥97%

Supplier: AOBChem USA

Warning

424041-5G 424041-1G 424041-25G 424041-500MG
77629-208EA 1484.23 USD
77629-208 77638-686 77659-258 77650-684
2-[(5-Chloro-2-pyridinyl)oxy]-2-methylpropanoic acid ≥97%
2-[(5-Chloro-2-pyridinyl)oxy]-2-methylpropanoic acid

Precaución: For Research Use Only

Formula: 215.63 g/mol
MW: C₉H₁₀ClNO₃
Storage Temperature: Ambient
CAS Number: 32230-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR