4-Bromo-α-methylbenzo[b]thiophene-2-methanol ≥95%

Supplier: AOBChem USA

Warning

423984-1G 423984-5G 423984-500MG
77638-684EA 637 USD
77638-684 77659-256 77646-732
4-Bromo-α-methylbenzo[b]thiophene-2-methanol ≥95%
4-Bromo-α-methylbenzo[b]thiophene-2-methanol

Precaución: For Research Use Only

Formula: 257.15 g/mol
MW: C₁₀H₉BrOS
Storage Temperature: Ambient
CAS Number: 1780551-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR