1-(4-Bromobenzo[b]thien-2-yl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

423969-1G 423969-5G 423969-500MG
77638-682EA 523 USD
77638-682 77655-672 77650-682
1-(4-Bromobenzo[b]thien-2-yl)-1-propanone ≥95%
1-(4-Bromobenzo[b]thien-2-yl)-1-propanone

Precaución: For Research Use Only

Formula: 269.16 g/mol
MW: C₁₁H₉BrOS
Storage Temperature: Ambient
CAS Number: 1217889-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR