4-Bromo-2-chlorobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

423967-5G 423967-1G 423967-500MG
77629-206EA 1484.23 USD
77629-206 77638-680 77650-680
4-Bromo-2-chlorobenzo[b]thiophene ≥95%
4-Bromo-2-chlorobenzo[b]thiophene

Precaución: For Research Use Only

Formula: 247.54 g/mol
MW: C₈H₄BrClS
Storage Temperature: Ambient
CAS Number: 445280-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR