4-Chloro-α,α-dimethylbenzo[b]thiophene-2-methanol ≥95%

Supplier: AOBChem USA

Warning

423917-1G 423917-5G 423917-500MG
77638-676EA 574 USD
77638-676 77659-252 77650-676
4-Chloro-α,α-dimethylbenzo[b]thiophene-2-methanol ≥95%
4-Chloro-α,α-dimethylbenzo[b]thiophene-2-methanol

Precaución: For Research Use Only

Formula: C₁₁H₁₁ClOS
Storage Temperature: Ambient
CAS Number: 1895962-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR