1-(4-Chlorobenzo[b]thien-2-yl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

423814-500MG 423814-5G 423814-1G
77652-792EA 414 USD
77652-792 77659-250 77642-714
1-(4-Chlorobenzo[b]thien-2-yl)-1-propanone ≥95%
1-(4-Chlorobenzo[b]thien-2-yl)-1-propanone

Precaución: For Research Use Only

Formula: 224.71 g/mol
MW: C₁₁H₉ClOS
Storage Temperature: Ambient
CAS Number: 360575-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR