1-(4-Chlorobenzo[b]thien-2-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

423807-5G 423807-1G 423807-25G 423807-500MG
77629-204EA 1484.23 USD
77629-204 77638-674 77655-670 77646-730
1-(4-Chlorobenzo[b]thien-2-yl)cyclobutanol ≥95%
1-(4-Chlorobenzo[b]thien-2-yl)cyclobutanol

Precaución: For Research Use Only

Formula: 238.73 g/mol
MW: C₁₂H₁₁ClOS
Storage Temperature: Ambient
CAS Number: 2762931-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR