2-Bromo-4-chlorobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

423797-5G 423797-1G 423797-25G 423797-500MG
77633-098EA 1417 USD
77633-098 77638-672 77655-668 77650-674
2-Bromo-4-chlorobenzo[b]thiophene ≥95%
2-Bromo-4-chlorobenzo[b]thiophene

Precaución: For Research Use Only

Formula: 247.54 g/mol
MW: C₈H₄BrClS
Storage Temperature: Ambient
CAS Number: 66490-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR