1-(4-Chlorobenzo[b]thien-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

423794-5G 423794-25G 423794-1G 423794-500MG
77633-096EA 753 USD
77633-096 77659-248 77642-712 77646-728
1-(4-Chlorobenzo[b]thien-2-yl)ethanone ≥95%
1-(4-Chlorobenzo[b]thien-2-yl)ethanone

Precaución: For Research Use Only

Formula: 210.68 g/mol
MW: C₁₀H₇ClOS
Storage Temperature: Ambient
CAS Number: 66490-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR