5-Bromo-2,2-difluoro-4-methoxybenzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

422673-5G 422673-25G 422673-500MG 422673-1G
77629-200EA 1484.23 USD
77629-200 77635-790 77652-788 77642-710
5-Bromo-2,2-difluoro-4-methoxybenzo[d][1,3]dioxole ≥95%
5-Bromo-2,2-difluoro-4-methoxybenzo[d][1,3]dioxole

Precaución: For Research Use Only

Formula: 267.0243 g/mol
MW: C₈H₅BrF₂O₃
Storage Temperature: Ambient
CAS Number: 1628572-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR