Cyclopropyl(2,3-dichloropyridin-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

422591-5G 422591-1G 422591-500MG
77629-190EA 1484.23 USD
77629-190 77642-704 77650-670
Cyclopropyl(2,3-dichloropyridin-4-yl)methanol ≥95%
Cyclopropyl(2,3-dichloropyridin-4-yl)methanol

Precaución: For Research Use Only

Formula: C₉H₉Cl₂NO
Storage Temperature: Ambient
CAS Number: 2354029-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR