4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanol, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol

42259-5G 42259-1G 42259-25G
77635-012EA 212 USD
77635-012 77638-664 77655-660
4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester ≥95%
4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18733422
CAS Number: 1173922-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR