Cyclobutyl(2,3-dichloropyridin-4-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

422577-5G 422577-1G 422577-500MG
77629-186EA 1484.23 USD
77629-186 77638-662 77650-666
Cyclobutyl(2,3-dichloropyridin-4-yl)methanone ≥95%
Cyclobutyl(2,3-dichloropyridin-4-yl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₉Cl₂NO
Storage Temperature: Ambient
CAS Number: 2354253-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR