2-(2,6-Difluoro-3-methoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

422554-5G 422554-25G 422554-1G 422554-500MG
77633-094EA 1357 USD
77633-094 77655-656 77642-700 77650-664
2-(2,6-Difluoro-3-methoxyphenyl)propan-2-ol ≥95%
2-(2,6-Difluoro-3-methoxyphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 202.198 g/mol
MW: C₁₀H₁₂F₂O₂
Storage Temperature: Ambient
CAS Number: 1891402-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR