(5-Chloro-2-methylphenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

422442-5G 422442-1G 422442-500MG
77655-654EA 2001 USD
77655-654 77642-696 77646-716
(5-Chloro-2-methylphenyl)(phenyl)methanone ≥95%
(5-Chloro-2-methylphenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 230.69 g/mol
MW: C₁₄H₁₁ClO
Storage Temperature: Ambient
CAS Number: 33184-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR