2-Bromo-5-(trimethylsilyl)thiophene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2-thienyl)trimethylsilane

421318-5G 421318-1G 421318-500MG 421318-25G
77635-010EA 490 USD
77635-010 77638-656 77652-782 77659-240
2-Bromo-5-(trimethylsilyl)thiophene ≥97%
2-Bromo-5-(trimethylsilyl)thiophene

Precaución: For Research Use Only

Formula: C₇H₁₁BrSSi
MW: 235.21 g/mol
Storage Temperature: Ambient
CAS Number: 18246-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR