2-Ethyl-1-(3-methoxy-5-methylphenyl)butan-1-ol ≥97%

Supplier: AOBChem USA

Warning

421299-5G 421299-25G 421299-1G 421299-500MG
77629-174EA 1484.23 USD
77629-174 77655-650 77642-692 77650-660
2-Ethyl-1-(3-methoxy-5-methylphenyl)butan-1-ol ≥97%
2-Ethyl-1-(3-methoxy-5-methylphenyl)butan-1-ol

Precaución: For Research Use Only

Formula: 222.3232 g/mol
MW: C₁₄H₂₂O₂
Storage Temperature: Ambient
CAS Number: 3037692-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR