2-(Tert-butoxy)-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

421259-5G 421259-25G 421259-1G 421259-500MG
77629-160EA 1484.23 USD
77629-160 77655-644 77642-686 77646-708
2-(Tert-butoxy)-4-iodopyridine ≥97%
2-(Tert-butoxy)-4-iodopyridine

Precaución: For Research Use Only

Formula: C₉H₁₂INO
Storage Temperature: Ambient
CAS Number: 3031788-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR