2-(Tert-butoxy)-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

421257-5G 421257-25G 421257-1G 421257-500MG
77635-008EA 370 USD
77635-008 77659-234 77642-684 77646-706
2-(Tert-butoxy)-3-iodopyridine ≥97%
2-(Tert-butoxy)-3-iodopyridine

Precaución: For Research Use Only

Formula: 277.1021 g/mol
MW: C₉H₁₂INO
Storage Temperature: Ambient
CAS Number: 1020253-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR