2-(Tert-butoxy)-3-chloropyridine ≥95%

Supplier: AOBChem USA

Warning

421255-5G 421255-25G 421255-1G 421255-500MG
77629-156EA 1777.78 USD
77629-156 77659-232 77642-682 77650-648
2-(Tert-butoxy)-3-chloropyridine ≥95%
2-(Tert-butoxy)-3-chloropyridine

Precaución: For Research Use Only

Formula: 185.6506 g/mol
MW: C₉H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 1289149-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR