6-Bromo-2-(tert-butoxy)-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

421249-5G 421249-1G 421249-500MG
77659-230EA 2168 USD
77659-230 77642-680 77646-704
6-Bromo-2-(tert-butoxy)-3-iodopyridine ≥97%
6-Bromo-2-(tert-butoxy)-3-iodopyridine

Precaución: For Research Use Only

Formula: 355.9982 g/mol
MW: C₉H₁₁BrINO
Storage Temperature: Ambient
CAS Number: 3037704-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR