(E)-1-bromo-4-(but-1-en-1-yl)benzene ≥97%

Supplier: AOBChem USA

Warning

421222-5G 421222-25G 421222-1G 421222-500MG
77629-154EA 1821.04 USD
77629-154 77655-642 77642-678 77646-702
(E)-1-bromo-4-(but-1-en-1-yl)benzene ≥97%
(E)-1-bromo-4-(but-1-en-1-yl)benzene

Precaución: For Research Use Only

Formula: 211.0983 g/mol
MW: C₁₀H₁₁Br
Storage Temperature: Ambient
CAS Number: 490035-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR