4-(2-Methylprop-1-en-1-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

421219-5G 421219-1G 421219-500MG
77629-152EA 1484.23 USD
77629-152 77638-642 77646-700
4-(2-Methylprop-1-en-1-yl)benzaldehyde ≥95%
4-(2-Methylprop-1-en-1-yl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₁₂O
Storage Temperature: Ambient
CAS Number: 145589-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR