1-(4-Chloro-3-isopropylphenyl)prop-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

421136-5G 421136-1G 421136-500MG
77659-228EA 2168 USD
77659-228 77642-674 77650-646
1-(4-Chloro-3-isopropylphenyl)prop-2-en-1-ol ≥97%
1-(4-Chloro-3-isopropylphenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 210.7 g/mol
MW: C₁₂H₁₅ClO
Storage Temperature: Ambient
CAS Number: 3031788-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR