(2-Chloro-4-methylphenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

420771-5G 420771-25G 420771-1G 420771-500MG
77633-086EA 1441 USD
77633-086 77659-220 77642-666 77650-642
(2-Chloro-4-methylphenyl)(phenyl)methanone ≥95%
(2-Chloro-4-methylphenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 230.69 g/mol
MW: C₁₄H₁₁ClO
Storage Temperature: Ambient
CAS Number: 71549-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR