3'-Chloro-2'-propoxy-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%

Supplier: AOBChem USA

Warning

420596-5G 420596-25G 420596-1G 420596-500MG
77629-142EA 1484.23 USD
77629-142 77659-218 77642-662 77650-638
3'-Chloro-2'-propoxy-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%
3'-Chloro-2'-propoxy-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol

Precaución: For Research Use Only

Formula: 266.763 g/mol
MW: C₁₅H₁₉ClO₂
Storage Temperature: Ambient
CAS Number: 3037696-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR