(3-Bromo-5-chloro-2-fluorophenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

420568-5G 420568-1G 420568-25G 420568-500MG
77633-084EA 1441 USD
77633-084 77638-634 77655-636 77646-690
(3-Bromo-5-chloro-2-fluorophenyl)(cyclopropyl)methanone ≥95%
(3-Bromo-5-chloro-2-fluorophenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: 277.517 g/mol
MW: C₁₀H₇BrClFO
Storage Temperature: Ambient
CAS Number: 3031792-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR