1-(3-Bromo-5-chloro-2-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

420566-5G 420566-25G 420566-1G 420566-500MG
77629-140EA 1484.23 USD
77629-140 77659-216 77642-660 77646-688
1-(3-Bromo-5-chloro-2-fluorophenyl)propan-1-one ≥95%
1-(3-Bromo-5-chloro-2-fluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: 265.507 g/mol
MW: C₉H₇BrClFO
Storage Temperature: Ambient
CAS Number: 3031805-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR