3-(Tert-butylthio)-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

420506-5G 420506-1G 420506-25G 420506-500MG
77629-138EA 1484.23 USD
77629-138 77638-632 77659-214 77650-636
3-(Tert-butylthio)-4-(trifluoromethyl)benzaldehyde ≥97%
3-(Tert-butylthio)-4-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₂H₁₃F₃OS
Storage Temperature: Ambient
CAS Number: 3031799-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR