3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3,5-Dimethyl-1-(tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
41885-5G
41885-1G
41885-25G
77635-002EA
180
USD
77635-002
77638-626
77659-208
3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester ≥97%
3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester
Precaución: For Research Use Only
Formula:
C₁₆H₂₇BN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD12031452 CAS Number: 1126779-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...