2-(Cyclopropylmethoxy)-4-trifluoromethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

41789-5G 41789-25G 41789-1G 41789-500MG
77635-000EA 436 USD
77635-000 77659-206 77642-658 77650-632
2-(Cyclopropylmethoxy)-4-trifluoromethylphenylboronic acid ≥95%
2-(Cyclopropylmethoxy)-4-trifluoromethylphenylboronic acid

Precaución: For Research Use Only

Formula: 260.017 g/mol
MW: C₁₁H₁₂BF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18087694
CAS Number: 2096336-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR