4-Bromo-1-methoxy-2,3-dihydro-1H-indene ≥97%
Supplier: AOBChem USA
|
Warning
|
415557-1G
415557-5G
415557-500MG
77638-622EA
574
USD
77638-622
77659-202
77646-684
4-Bromo-1-methoxy-2,3-dihydro-1H-indene ≥97%
4-Bromo-1-methoxy-2,3-dihydro-1H-indene
Precaución: For Research Use Only
Formula:
227.0977 g/mol MW: C₁₀H₁₁BrO Storage Temperature: Ambient |
CAS Number:
858224-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...