(3-(Benzyloxy)-2,6-difluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

415361-5G 415361-1G 415361-25G 415361-500MG
77633-080EA 1441 USD
77633-080 77638-616 77655-624 77650-618
(3-(Benzyloxy)-2,6-difluorophenyl)trimethylsilane ≥95%
(3-(Benzyloxy)-2,6-difluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 292.396 g/mol
MW: C₁₆H₁₈F₂OSi
Storage Temperature: Ambient
CAS Number: 152434-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR