1-(5-Chloro-4-methylthiophen-2-yl)-2,2-difluoropropan-1-one ≥97%

Supplier: AOBChem USA

Warning

415038-1G 415038-5G 415038-500MG
77638-610EA 592 USD
77638-610 77655-614 77650-612
1-(5-Chloro-4-methylthiophen-2-yl)-2,2-difluoropropan-1-one ≥97%
1-(5-Chloro-4-methylthiophen-2-yl)-2,2-difluoropropan-1-one

Precaución: For Research Use Only

Formula: 224.655 g/mol
MW: C₈H₇ClF₂OS
Storage Temperature: Ambient
CAS Number: 1878049-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR